Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ovca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027136.3 | NP_081412.1 | 2 | 1080 | ||
GenBank mRNA | AK030338.1 | - | 2 | 1082 | ||
AK033511.1 | - | 2 | 1080 | |||
AK049783.1 | - | 1 | 1332 | |||
BC056976.1 | - | 2 | 1080 | |||
BC060159.1 | - | 2 | 1080 |
Gene Symbol: Dph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128996.1 | - | 2232 |