Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081406.1 | NP_001074875.1 | 1, 2 | 257 | ||
GenBank mRNA | AK010419.1 | - | 1, 2 | 250 | ||
AK144741.1 | - | 1, 2 | 258 | |||
BC022648.1 | - | 1, 2 | 211 | |||
BC075711.1 | - | 1, 2 | 239 | |||
BC117053.1 | - | 1, 2 | 236 | |||
BC117055.1 | - | 1, 2 | 236 |