Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168516.1 | NP_001161988.1 | 2 | 159 | ||
NM_001168517.1 | NP_001161989.1 | 2 | 206 | |||
NM_027476.3 | NP_081752.2 | 2 | 452 | |||
GenBank mRNA | AK017737.1 | - | 2 | 417 | ||
AK028164.1 | - | 2 | 415 | |||
AK043923.1 | - | 2 | 409 | |||
AK169337.1 | - | 2 | 411 | |||
AK170706.1 | - | 2 | 436 | |||
AK171568.1 | - | 2 | 436 | |||
BC056621.1 | - | 2 | 174 | |||
BC071194.1 | - | 2 | 393 | |||
CA559443.1 | - | 2 | 159 | |||
CD354313.1 | - | 2 | 115 |