Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brms1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037756.2 | NP_001032845.1 | 6 | 698 | ||
GenBank mRNA | AK003055.1 | - | 6 | 447 | ||
AK154678.1 | - | 6 | 699 | |||
AK155742.1 | - | 6 | 699 | |||
BC034885.1 | - | 5 | 562 | 1362264 |