Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285845.1 | NP_001272774.1 | 3 | 1326 | ||
NM_001285846.1 | NP_001272775.1 | 4 | 1252 | |||
NM_001285847.1 | NP_001272776.1 | 3 | 1215 | |||
NM_001285849.1 | NP_001272778.1 | 3 | 1099 | |||
NM_027995.3 | NP_082271.1 | 2 | 1131 | |||
GenBank mRNA | AF313618.1 | - | 2 | 885 | ||
AK009450.1 | - | 2 | 916 | |||
AK078685.1 | - | 2 | 889 | |||
AK161542.1 | - | 3 | 973 | |||
AK170307.1 | - | 4 | 1036 | |||
AK203036.1 | - | 1 | 33 | |||
AY278956.1 | - | 1 | 824 | |||
AY424296.1 | - | 1 | 860 | |||
BC022922.1 | - | 1 | 860 | 4239768 |
Set Membership: |
G-protein coupled receptor |