Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028049.2 | NP_082325.2 | 6 | 853 | ||
GenBank mRNA | AK002745.1 | - | 6 | 733 | ||
AK005473.1 | - | 5 | 681 | |||
AK179843.1 | - | 1 | 54 | |||
AK187855.1 | - | 1 | 57 | |||
AK188724.1 | - | 2 | 288 | |||
AK188900.1 | - | 1 | 57 | |||
AK189126.1 | - | 2 | 191 | |||
AK207864.1 | - | 2 | 291 | |||
AK211102.1 | - | 2 | 155 | |||
AK213133.1 | - | 2 | 288 | |||
AK219248.1 | - | 2 | 288 | |||
BC018273.1 | - | 6 | 793 | 4458296 |