Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028060.3 | NP_082336.3 | 8 | 1868 | ||
GenBank mRNA | AK005182.1 | - | 8 | 1664 | ||
BC019202.1 | - | 8 | 1670 | |||
BC050202.1 | - | 2 | 993 | |||
BC055843.1 | - | 8 | 1875 | 3991126 |