Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK154555.1 | - | 1 | 2051 | ||
AK155115.1 | - | 2 | 1970 | |||
BC026886.1 | - | 1 | 1883 | |||
BC043700.1 | - | 1 | 2035 | 5346204 |