Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028390.3 | NP_082666.2 | 24 | 3700 | ||
GenBank mRNA | AK045703.1 | - | 21 | 2664 | ||
AK046102.1 | - | 13 | 1486 | |||
BC030489.1 | - | 17 | 2042 | 5373071 | ||
BC032164.1 | - | 24 | 3718 | 5346703 |