Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028597.3 | NP_082873.2 | 3 | 640 | ||
GenBank mRNA | AK010668.1 | - | 3 | 620 | ||
BC019603.1 | - | 3 | 614 | 2647864 |