Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF061812.1 | - | 2, 3 | 630 | ||
AK290853.1 | - | 2, 3 | 693 | |||
S79867.1 | - | 2, 3 | 616 |