Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028790.3 | NP_083066.1 | 15 | 2249 | ||
GenBank mRNA | AK004905.1 | - | 15 | 2250 | ||
AK034622.1 | - | 15 | 2253 | |||
BC025852.1 | - | 15 | 2202 | |||
BC132050.1 | - | 15 | 2280 | 40130327 | ||
BC137996.1 | - | 15 | 2280 | 8861015 |