Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029211.2 | NP_083487.2 | 8, 9 | 860 | ||
GenBank mRNA | AK016037.1 | - | 8, 9 | 1091 | ||
AK034806.1 | - | 8, 9 | 2174 | |||
AK078509.1 | - | 8, 9 | 821 | |||
AK159032.1 | - | 8, 9 | 997 | |||
AK159260.1 | - | 8, 9 | 887 | |||
AK168821.1 | - | 8, 9 | 873 | |||
AK169089.1 | - | 8, 9 | 873 | |||
BC022686.1 | - | 8, 9 | 860 | 4241984 |