Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029494.2 | NP_083770.2 | 5 | 571 | ||
GenBank mRNA | AB232626.1 | - | 4 | 434 | ||
AF399756.1 | - | 4 | 434 | |||
AK017185.1 | - | 4 | 521 | |||
AK142206.1 | - | 5 | 578 | |||
BC017550.1 | - | 5 | 571 | 4503909 |