Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tusc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030254.3 | NP_084530.3 | 516 | |||
GenBank mRNA | AK028182.1 | - | 492 | |||
AK088616.1 | - | 530 | ||||
AK132766.1 | - | 539 | ||||
AK168664.1 | - | 474 | ||||
AK168961.1 | - | 474 | ||||
BC069898.1 | - | 516 | 6489807 |