Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204134.1 | NP_001191063.1 | 6 | 1162 | ||
NM_030888.4 | NP_112150.1 | 6 | 968 | |||
GenBank mRNA | AF246265.1 | - | 6 | 945 | ||
AK029352.1 | - | 6 | 945 | |||
AK075867.1 | - | 6 | 945 | |||
AK081704.1 | - | 6 | 945 | |||
AK128898.1 | - | 6 | 968 | |||
BC019636.1 | - | 6 | 1171 | |||
BC055047.1 | - | 6 | 930 | 6486507 | ||
BC055859.1 | - | 6 | 909 | 4238617 |