Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cadm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053199.3 | NP_444429.1 | 2, 3 | 432 | ||
GenBank mRNA | AF195662.1 | - | 2, 3 | 376 | ||
AK036352.1 | - | 3, 4 | 488 | |||
AK038917.1 | - | 2, 3 | 389 | |||
AK053077.1 | - | 2, 3 | 386 | |||
AY059393.1 | - | 2, 3 | 231 | |||
BC079837.1 | - | 3, 4 | 472 | |||
BC115417.2 | - | 2, 3 | 337 | 40093410 | ||
BC115418.2 | - | 2, 3 | 337 | 40093417 |