Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080633.2 | NP_542364.1 | 15 | 1943 | ||
GenBank mRNA | AK143917.1 | - | 15 | 1922 | ||
AK144207.1 | - | 15 | 1906 | |||
AK145511.1 | - | 15 | 1923 | |||
AK150027.1 | - | 15 | 1923 | |||
AK150954.1 | - | 16 | 1905 | |||
AK165411.1 | - | 15 | 1907 | |||
AK220091.1 | - | 2 | 223 | |||
BC004645.1 | - | 15 | 1969 | 3158239 | ||
BC094462.1 | - | 15 | 1902 | 4238599 | ||
CO429053.1 | - | 3 | 334 |