Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Socs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080843.2 | NP_543119.2 | 2 | 1091 | ||
GenBank mRNA | AK014115.1 | - | 2 | 1121 | ||
AY046322.1 | - | 1 | 919 | |||
BC117814.1 | - | 1 | 1012 | 40092681 | ||
BC117815.1 | - | 1 | 1012 | 40092683 |