Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146057.1 | NP_001139529.1 | 8 | 977 | ||
NM_001146058.1 | NP_001139530.1 | 8 | 839 | |||
NM_133348.2 | NP_579926.2 | 8 | 966 | |||
GenBank mRNA | AB049821.1 | - | 8 | 932 | ||
AB088411.1 | - | 8 | 905 | |||
AB088412.1 | - | 8 | 873 | |||
AB207243.1 | - | 8 | 932 | |||
AK010646.1 | - | 796 | ||||
AK152277.1 | - | 9 | 847 | |||
AK192638.1 | - | 1 | 20 | |||
BC013507.1 | - | 8 | 834 | 4224774 |