Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133683.3 | NP_598444.1 | 7 | 2093 | ||
GenBank mRNA | AK034298.1 | - | 7 | 2100 | ||
AK036228.1 | - | 6 | 2170 | |||
AK042426.1 | - | 6 | 2148 | |||
AK136778.1 | - | 2083 | ||||
AK138931.1 | - | 7 | 2081 | |||
AK155275.1 | - | 7 | 2054 | |||
AK170143.1 | - | 7 | 2053 | |||
AK171087.1 | - | 7 | 2053 | |||
AK189585.1 | - | 1 | 285 | |||
BC016895.1 | - | 7 | 2096 | 4239007 |