Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxxc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133687.2 | NP_598448.1 | 2 | 1153 | ||
GenBank mRNA | AK005364.1 | - | 2 | 1181 | ||
AK015150.1 | - | 2 | 1056 | |||
AK132950.1 | - | 2 | 1077 | |||
BC016207.1 | - | 1 | 844 | 4912395 | ||
BC089314.1 | - | 2 | 1153 | 30541251 |