Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133838.4 | NP_598599.2 | 1, 2 | 335 | ||
GenBank mRNA | AB239915.1 | - | 1, 2 | 246 | ||
AB239916.1 | - | 1, 2 | 246 | |||
AF173639.1 | - | 1, 2 | 251 | |||
AK147186.1 | - | 1, 2 | 287 | |||
AK149760.1 | - | 1, 2 | 279 | |||
AK159235.1 | - | 1, 2 | 295 | |||
AK159597.1 | - | 1, 2 | 295 | |||
AK163835.1 | - | 1, 2 | 279 | |||
AK166101.1 | - | 1, 2 | 346 | |||
AK169343.1 | - | 1, 2 | 346 | |||
BC007480.1 | - | 1, 2 | 375 | 3493227 | ||
BC012272.1 | - | 1, 2 | 268 | 3485836 |