Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195390.1 | NP_001182319.1 | 6 | 503 | ||
NM_133850.2 | NP_598611.2 | 5 | 428 | |||
GenBank mRNA | AK008447.1 | - | 5 | 383 | ||
AK009274.1 | - | 5 | 403 | |||
AK046582.1 | - | 5 | 378 | |||
AK077349.1 | - | 5 | 369 | |||
AK078774.1 | - | 5 | 387 | |||
AK089926.1 | - | 5 | 371 | |||
AK164805.1 | - | 5 | 337 | |||
AK169037.1 | - | 6 | 450 | |||
AK217917.1 | - | 2 | 48 | |||
BC003310.1 | - | 5 | 361 | |||
BC065794.1 | - | 5 | 329 | |||
BC092215.1 | - | 5 | 345 | 6819383 | ||
BC094216.1 | - | 5 | 385 | 6820900 |