Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133902.2 | NP_598663.2 | 5 | 450 | ||
GenBank mRNA | AF328927.1 | - | 6 | 717 | ||
AK014487.1 | - | 2 | 66 | |||
BC022601.1 | - | 5 | 450 | 4160199 |