Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133940.3 | NP_598701.2 | 1 | 2898 | ||
GenBank mRNA | AK036124.1 | - | 1 | 2795 | ||
AK084506.1 | - | 1 | 2785 | |||
AK155154.1 | - | 1 | 2770 | |||
BC006913.1 | - | 1 | 2475 | |||
BC021329.1 | - | 1 | 2779 | 3710945 |