Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133940.3 | NP_598701.2 | 1 | 2814 | ||
GenBank mRNA | AK036124.1 | - | 1 | 2711 | ||
AK084506.1 | - | 1 | 2701 | |||
AK155154.1 | - | 1 | 2686 | |||
BC006913.1 | - | 1 | 2391 | |||
BC021329.1 | - | 1 | 2695 | 3710945 |