Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286030.1 | NP_001272959.1 | 2 | 672 | ||
NM_001286031.1 | NP_001272960.1 | 2 | 541 | |||
NM_133941.2 | NP_598702.1 | 2 | 625 | |||
GenBank mRNA | AK028910.1 | - | 2 | 482 | ||
AK033624.1 | - | 2 | 627 | |||
AK135760.1 | - | 2 | 673 | |||
AK167889.1 | - | 2 | 570 | |||
AK169005.1 | - | 2 | 616 | |||
AK172504.1 | - | 2 | 606 | |||
AK204739.1 | - | 1 | 200 | |||
AY065980.1 | - | 2 | 627 | |||
BC006911.1 | - | 1 | 182 | |||
BC022920.1 | - | 1 | 381 | 4460064 |