Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snapc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133968.1 | NP_598729.1 | 5 | 983 | ||
GenBank mRNA | AK004379.1 | - | 5 | 983 | ||
AK035340.1 | - | 5 | 995 | |||
AK075569.1 | - | 5 | 1006 | |||
AK141475.1 | - | 5 | 987 | |||
AK146218.1 | - | 5 | 975 | |||
AK155012.1 | - | 5 | 664 | |||
AK181477.1 | - | 1 | 28 | |||
AK186997.1 | - | 1 | 19 | |||
BC011147.1 | - | 5 | 983 | 4194905 |