Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snapc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133968.1 | NP_598729.1 | 5 | 1291 | ||
GenBank mRNA | AK004379.1 | - | 5 | 1291 | ||
AK035340.1 | - | 5 | 1303 | |||
AK075569.1 | - | 5 | 1314 | |||
AK141475.1 | - | 5 | 1295 | |||
AK146218.1 | - | 5 | 1283 | |||
AK155012.1 | - | 5 | 972 | |||
AK181477.1 | - | 1 | 336 | |||
AK186997.1 | - | 1 | 327 | |||
AK188996.1 | - | 1 | 309 | |||
AK206395.1 | - | 1 | 229 | |||
AK211626.1 | - | 1 | 286 | |||
AK217227.1 | - | 1 | 296 | |||
BC011147.1 | - | 5 | 1291 | 4194905 |