Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164743.1 | NP_001158215.1 | 2 | 374 | ||
NM_134069.3 | NP_598830.2 | 3 | 498 | |||
GenBank mRNA | AA986956.1 | - | 1 | 25 | ||
AK128896.1 | - | 3 | 482 | |||
AK143896.1 | - | 3 | 470 | |||
AK165505.1 | - | 3 | 343 | |||
BC018252.1 | - | 3 | 427 | 4240254 | ||
BC024459.1 | - | 3 | 454 | 4975341 | ||
BC034179.1 | - | 3 | 318 | 4972500 | ||
BC034183.1 | - | 3 | 303 | 4973620 |