Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243041.1 | NP_001229970.1 | 9 | 963 | ||
NM_134079.4 | NP_598840.1 | 9 | 920 | |||
GenBank mRNA | AK050135.1 | - | 3 | 223 | ||
AK151863.1 | - | 9 | 965 | |||
AK160264.1 | - | 9 | 917 | |||
AY540996.1 | - | 9 | 862 | |||
AY540997.1 | - | 8 | 807 | |||
BC009659.1 | - | 9 | 885 | 3485959 |