Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134091.2 | NP_598852.2 | 5 | 449 | ||
GenBank mRNA | AK142314.1 | - | 5 | 418 | ||
AK170806.1 | - | 5 | 416 | |||
AK185066.1 | - | 1 | 34 | |||
AK189808.1 | - | 1 | 34 | |||
AK206238.1 | - | 1 | 34 | |||
AY382616.1 | - | 5 | 403 | |||
BC018197.1 | - | 5 | 409 | 4190407 | ||
BC024122.1 | - | 5 | 409 | 5097970 | ||
BC025561.1 | - | 5 | 387 | 5322980 | ||
BY242851.1 | - | 5 | 451 |