Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncald | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170866.1 | NP_001164337.1 | 7 | 2897 | ||
NM_001170867.1 | NP_001164338.1 | 5 | 2831 | |||
NM_001170868.1 | NP_001164339.1 | 4 | 2779 | |||
NM_134094.4 | NP_598855.2 | 7 | 3087 | |||
GenBank mRNA | AK034031.1 | - | 5 | 2810 | ||
AK048440.1 | - | 7 | 3068 | |||
AK085416.1 | - | 1 | 1435 | |||
AK167313.1 | - | 4 | 2747 | |||
BC011162.1 | - | 4 | 2774 | 4191195 | ||
BC026979.1 | - | 4 | 2733 | 5365560 |