Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPDL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207000.1 | NP_001193929.1 | 1, 2 | 978 | ||
NM_031372.3 | NP_112740.1 | 1, 2 | 978 | |||
NR_003249.2 | 1, 2 | 978 | ||||
GenBank mRNA | AB017019.1 | - | 1, 2 | 978 | ||
AB066484.1 | - | 1, 2 | 86 | |||
AK300122.1 | - | 1, 2 | 64 | |||
AY453824.1 | - | 1, 2 | 77 | |||
BC007392.2 | - | 1, 2 | 430 | |||
BC011714.2 | - | 1, 2 | 619 | 3929324 | ||
BC018961.2 | - | 1, 2 | 619 | |||
BC071944.1 | - | 1, 2 | 559 | 5015862 | ||
CR407623.1 | - | 1, 2 | 443 | |||
D89092.1 | - | 1, 2 | 194 | |||
D89678.1 | - | 1, 2 | 53 |