Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3il1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144538.2 | NP_653121.1 | 9 | 1157 | ||
GenBank mRNA | AK004767.1 | - | 8 | 933 | ||
AK132764.1 | - | 12 | 1864 | |||
AK161298.1 | - | 12 | 1864 | |||
BC020147.1 | - | 9 | 1122 | 3708973 |