Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172095.1 | NP_001165566.1 | 6 | 817 | ||
NM_144787.2 | NP_659036.1 | 6 | 881 | |||
GenBank mRNA | AK042792.1 | - | 5 | 596 | ||
AK048092.1 | - | 6 | 794 | |||
AK053104.1 | - | 6 | 757 | |||
AK081769.1 | - | 6 | 824 | |||
AK144103.1 | - | 6 | 844 | |||
AK144890.1 | - | 6 | 855 | |||
AK173023.1 | - | 6 | 881 | |||
BC020180.1 | - | 6 | 823 | 4923943 | ||
BC042424.1 | - | 6 | 817 | 5357211 |