Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033597.1 | NP_001028769.1 | 13 | 1372 | ||
NM_001033598.1 | NP_001028770.1 | 13 | 1341 | |||
NM_001033599.1 | NP_001028771.1 | 13 | 1547 | |||
NM_144823.4 | NP_659072.3 | 13 | 1372 | |||
GenBank mRNA | AB073974.1 | - | 12 | 1216 | ||
AK039380.1 | - | 13 | 1458 | |||
AK122384.1 | - | 2 | 3138 | |||
AK147283.1 | - | 13 | 1342 | |||
AK147421.1 | - | 13 | 1373 | |||
AY167035.1 | - | 13 | 1342 | |||
AY786360.1 | - | 15 | 1890 | |||
AY786361.1 | - | 13 | 1322 | |||
AY786362.1 | - | 13 | 1514 | |||
AY786363.1 | - | 13 | 1367 | |||
BC016114.1 | - | 13 | 1298 | 4481949 | ||
BC022959.1 | - | 13 | 1372 | 5362660 |