Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uhrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144873.2 | NP_659122.2 | 3 | 737 | ||
GenBank mRNA | AB116653.1 | - | 3 | 472 | ||
AF274047.1 | - | 3 | 647 | |||
AK031036.1 | - | 3 | 799 | |||
AK041564.1 | - | 3 | 759 | |||
AK042321.1 | - | 3 | 778 | |||
AK051743.1 | - | 3 | 778 | |||
AK080925.1 | - | 3 | 778 | |||
AK172682.1 | - | 3 | 621 | |||
AK206406.1 | - | 2 | 183 | |||
BC060241.1 | - | 3 | 737 | 6334856 |