Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144948.5 | NP_659197.2 | 5 | 1771 | ||
GenBank mRNA | AA178228.1 | - | 1 | 428 | ||
AF458961.1 | - | 5 | 1735 | |||
AK005200.1 | - | 5 | 1747 | |||
AK008645.1 | - | 5 | 1730 | |||
AK145381.1 | - | 4 | 2594 | |||
AK151056.1 | - | 5 | 1750 | |||
AK151827.1 | - | 5 | 1770 | |||
AK153504.1 | - | 6 | 1840 | |||
BC054774.1 | - | 5 | 1738 | 6807637 |