Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pxk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145458.3 | NP_663433.2 | 1, 2 | 211 | ||
NM_178279.2 | NP_840063.1 | 1, 2 | 211 | |||
GenBank mRNA | AK028501.1 | - | 1, 2 | 160 | ||
AK048910.1 | - | 1, 2 | 171 | |||
AK147037.1 | - | 1, 2 | 212 | |||
AK150337.1 | - | 1, 2 | 196 | |||
AK150413.1 | - | 1, 2 | 196 | |||
AK150660.1 | - | 1, 2 | 212 | |||
AK151098.1 | - | 1, 2 | 170 | |||
AK152058.1 | - | 1, 2 | 170 | |||
AK152789.1 | - | 1, 2 | 170 | |||
AK153067.1 | - | 1, 2 | 170 | |||
AK156146.1 | - | 1, 2 | 177 | |||
AK157364.1 | - | 1, 2 | 205 | |||
AK166036.1 | - | 1, 2 | 136 | |||
AK169112.1 | - | 1, 2 | 171 | |||
AK169565.1 | - | 1, 2 | 172 | |||
AK170361.1 | - | 1, 2 | 207 | |||
AK172347.1 | - | 1, 2 | 160 | |||
AK172463.1 | - | 1, 2 | 212 | |||
BC016131.1 | - | 1, 2 | 181 | 4922274 | ||
BY168060.1 | - | 1, 2 | 209 | |||
DQ095196.1 | - | 1, 2 | 109 | |||
DQ095197.1 | - | 1, 2 | 109 |