Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Deptor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037937.3 | NP_001033026.1 | 2 | 254 | ||
NM_145470.3 | NP_663445.2 | 2 | 432 | |||
GenBank mRNA | AK139888.1 | - | 2 | 254 | ||
AK145208.1 | - | 2 | 326 | |||
AK184075.1 | - | 1 | 133 | |||
AK220300.1 | - | 2 | 266 | |||
BC004774.1 | - | 2 | 437 | |||
DV649395.1 | - | 2 | 399 |