Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145620.4 | NP_663595.1 | 6 | 457 | ||
GenBank mRNA | AF368232.1 | - | 6 | 440 | ||
AK076113.1 | - | 6 | 459 | |||
AK156199.1 | - | 6 | 449 | |||
BC014703.1 | - | 6 | 457 | 4237895 |