Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF411211.1 | - | 2 | 336 | ||
AF411212.1 | - | 4 | 675 | |||
BC013080.1 | - | 4 | 644 | 3499860 | ||
BC037233.1 | - | 4 | 696 | 4951512 |