Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161406.1 | NP_001154878.1 | 16 | 2129 | ||
NM_145890.2 | NP_665897.2 | 14 | 1761 | |||
GenBank mRNA | AF411211.1 | - | 14 | 1761 | ||
AF411212.1 | - | 16 | 2100 | |||
AK076307.1 | - | 16 | 2115 | |||
AK161048.1 | - | 16 | 2011 | |||
BC013080.1 | - | 16 | 2069 | 3499860 | ||
BC037233.1 | - | 16 | 2121 | 4951512 |