Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005481.2 | NP_005472.2 | 11 | 1941 | ||
XM_017026120.1 | XP_016881609.1 | 10 | 1729 | |||
XM_017026121.1 | XP_016881610.1 | 10 | 1781 | |||
GenBank mRNA | AF106934.1 | - | 11 | 1928 | ||
AF121228.1 | - | 10 | 1790 | |||
AL137732.1 | - | 4 | 643 | |||
BC004554.1 | - | 11 | 1922 | 4053743 | ||
BC007853.2 | - | 7 | 1051 | |||
BC011841.1 | - | 11 | 1922 | 4053664 | ||
BC017282.2 | - | 11 | 1916 | 4876977 | ||
EU392449.1 | - | 9 | 1600 | |||
EU392451.1 | - | 9 | 1369 | |||
EU392452.1 | - | 7 | 1090 | |||
EU392453.1 | - | 7 | 1084 | |||
EU392454.1 | - | 7 | 961 | |||
EU392455.1 | - | 5 | 526 | |||
EU392456.1 | - | 5 | 571 | |||
EU392457.1 | - | 4 | 484 | |||
EU392458.1 | - | 4 | 400 | |||
EU392459.1 | - | 4 | 391 | |||
EU392460.1 | - | 3 | 280 | |||
EU392461.1 | - | 3 | 130 | |||
EU392462.1 | - | 2 | 249 | |||
EU392463.1 | - | 2 | 148 | |||
EU392484.1 | - | 6 | 716 | |||
EU392485.1 | - | 6 | 555 | |||
EU392486.1 | - | 5 | 414 | |||
EU392487.1 | - | 5 | 626 | |||
EU392488.1 | - | 5 | 359 | |||
EU392489.1 | - | 5 | 485 | |||
EU392490.1 | - | 4 | 284 | |||
EU392491.1 | - | 3 | 165 | |||
EU392492.1 | - | 4 | 248 | |||
EU392493.1 | - | 2 | 254 | |||
EU392494.1 | - | 3 | 332 | |||
EU392495.1 | - | 2 | 255 | |||
EU392496.1 | - | 4 | 198 |