Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190343.1 | NP_001177272.1 | 9 | 1614 | ||
NM_001190344.1 | NP_001177273.1 | 10 | 1700 | |||
NM_146014.3 | NP_666126.1 | 10 | 1788 | |||
GenBank mRNA | AK155145.1 | - | 10 | 1762 | ||
AK166131.1 | - | 7 | 1315 | |||
AK169727.1 | - | 10 | 1703 | |||
BC029157.1 | - | 10 | 1719 | 4952288 |