Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005484.3 | NP_005475.2 | 2, 3 | 269 | ||
XM_005267247.3 | XP_005267304.1 | 2, 3 | 269 | |||
GenBank mRNA | AF085734.1 | - | 2, 3 | 266 | ||
AJ236876.2 | - | 1, 2 | 180 | |||
AK001980.1 | - | 2, 3 | 246 | |||
DA354586.1 | - | 2, 3 | 269 |