Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6kl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146244.4 | NP_666356.2 | 5, 6 | 648 | ||
GenBank mRNA | AK031847.1 | - | 5, 6 | 639 | ||
AK044050.1 | - | 5, 6 | 639 | |||
BC027298.1 | - | 5, 6 | 602 | 3672127 |