Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146252.2 | NP_666364.1 | 6, 7 | 413 | ||
GenBank mRNA | AK158066.1 | - | 6, 7 | 494 | ||
BC025586.1 | - | 6, 7 | 439 | 5324875 |